Improve quality control and ensure safe operation in the coal mine
From the mine to loadout, we’ve got everything you need to meet quality specifications and ship a more consistent, quality product. Thermo Scientific online elemental coal analyzers and blending software provide coal producers with accurate, reliable data to control coal blends and ensure on-spec batches.
We also offer belt scales, weigh feeders, and coal quality analysis and coal sampling units that provide the process information needed to manage your operation to its greatest efficiency while giving you and your customers the assurance that you are shipping—and they are receiving—the tonnages required.
Create a safer environment for your workers and protect your equipment with our tramp metal detectors, level sensors, belt misalignment switches, emergency pull switches, plugged chute switches, and personal dust monitors.
Thermo Fisher Scientific has served the coal industry for over 35 years
This animation shows a typical coal processing plant where coal is taken from the mine pit to the processing plant and then through the coal manufacturing process. It includes multiple products tailored to the application.
Related coal processing products
Oretronic IV Tramp Metal Detector
Protect expensive crushers, conveyors and other process equipment from damage by tramp metal. Designed for belt conveyors moving coal, iron pellets, minerals, aggregates and other bulk materials, Oretronic Tramp Metal Detectors detect all types of metallic scrap that conventional metal detectors may miss, including bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains and tools. It can even detect tramp metal when buried in wet conductive materials.
Ramsey Series 14 Precision Belt Scale System
Achieve high accuracy in your weighing applications requiring certification by government and regulatory agencies using the Thermo Scientific™ Ramsey™ Series 14 Belt Scale System. The Ramsey Series 14 Belt Scale System consists of a 4-idler, 4-load cell weighbridge, speed sensor and electronics to provide stable high-accuracy weighing. NMI & NTEP certification and accuracy of +/- 0.125% of rated capacity for 10-14-4 scales.
Pro-Line Conveyor Protection Switches
Protect your equipment, prevent accidents and reduce unscheduled shutdowns with the Thermo Scientific™ Pro-Line Conveyor Protection Switches. These conveyor protection switches are used for position information, control signals, and to identify potentially hazardous situations with your process equipment. When hazardous situations occur they activate alarms to stop your equipment. This keeps your personnel safe and limits damage to your equipment. Numerous options allow you to pick the right switch to fit your applications. Use the conveyor protection switches to protect your business in even the most challenging applications.
ECA-3 Online Elemental Coal Analyzer
Provides real-time quality analysis of critical process streams to facilitate sorting, blending and out-of-seam dilution control. Productivity is improved in mine management, preparation plants and inventory management. The ECA-3 Analyzer is ideal for raw coal, plant feed, clean coal and blended coal applications. Coal fired power plants benefit by reducing the use of higher, more expensive quality coals while increasing boiler efficiency.
CQM FLEX Coal Analyzer
The CQM Flex can incorporate an electrically-driven neutron generator, or Californium (252Cf) as its source of neutrons to meet the needs of various application requirements. The CQM FLEX measures and reports levels of sulfur, moisture, total ash, calorific value, elemental ash composition, and other critical parameters required by both coal consumers and producers to maximize the use of coal resources and improve efficiencies.
DDG3 Digital Density Gauge
With the latest controller and communication protocols options, this non-intrusive, non-contact nuclear density gauge is suitable for applications containing a high temperature, highly toxic or highly corrosive process and helps to ensure accurate process control and optimum productivity in industries including mining processing, petrochemical, coal operations, food processing, water treatment, dredging, and pulp and paper.
RadEye™ NL Personal Highly Sensitive Neutron Radiation Detectors
The new RadEye N / NL closes a gap in the classical product spectrum of the radiation measurement technology. Rem-Counters for neutron detection with a He-3 or BF3 tube are usually heavy and bulky since fast neutrons have to be moderated in order to be detected and to provide the correct neutron dose rate response. Low energy neutrons with their lower biological impact however have to be suppressed to a large extent. Dropping the demand for energy compensated dose rate response, a device can be built with one fiftieth of the weight and comparable or even a higher neutron response as compared to a Rem-Counter.
Speak with our solution specialist
Please complete the form below to request for a sales representative to contact you.
Email Address*
First Name*
Last Name*
Company*
Job Title*
Location*
Business Phone*
Select the solution of your interest
I would like to receive marketing and promotional messages about Thermo Scientific products and services.
Additional Resources
PDF  Brochure: Instrument Solutions for Coal Mining Industry »
PDF  Specification Sheet: Oretonic IV Tramp Metal Detector »
PDF  Specification Sheet: Ramsey Series 14 Belt Scale System » 
PDF  Specification Sheet: Ramsey Conveyor Protection Switches » 
PDF  Specification Sheet: ECA-3 Elemental Crossbelt Analyzer »
PDF  Specification Sheet: CQM Flex Coal Analyzer »
PDF  Specification Sheet: DDG3 Digital Density Gauge »
PDF  Specification Sheet: RadEye NL Personal Highly Sensitive Neutron Radiation Detectors »
Coal Blog
Stay at the forefront of the Mining Industry! Get to know the newest technological advances in our blog Advancing Mining - Coal